CAS RN 52450-38-1
CAS RN 52450-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Gamma-Acetyl-N-2-formyl-5-methoxykynurenamine, TRC
CAS: 52450-38-1 Molecular Formula: C13 H16 N2 O4 Molecular Weight (g/mol): 264.28 Synonym: N-[3-[2-(Formylamino)-5-methoxyphenyl]-3-oxopropyl]acetamide,N1-Acetyl-N2-formyl-5-methoxykynuramine,NSC 688263 IUPAC Name: N-[3-(2-formamido-5-methoxyphenyl)-3-oxopropyl]acetamide SMILES: COc1ccc(NC=O)c(c1)C(=O)CCNC(=O)C