accessibility menu, dialog, popup

UserName

H3COOClO

CAS RN 5205-39-0

IUPAC Name: ethyl 5-chloro-5-oxopentanoate
Synonyms: ethyl 5-chloro-5-oxopentanoate
Molecular Weight (g/mol): 178.61
Molecular Formula: C7H11ClO3
InChi Key: KKJAQUGGQMCNJY-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCC(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)