accessibility menu, dialog, popup

UserName

NSClOHO

CAS RN 5198-87-8

IUPAC Name: 2-chloro-1,3-thiazole-4-carboxylic acid
Synonyms: 2-chloro-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 163.58
Molecular Formula: C4H2ClNO2S
InChi Key: UVYJJJQMZPCYKY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(Cl)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results