CAS RN 51940-44-4
IUPAC Name:
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
Synonyms:
pipemidic acid 吡哌酸
Molecular Weight (g/mol):
303.32
Molecular Formula:
C14H17N5O3
InChi Key:
JOHZPMXAZQZXHR-UHFFFAOYSA-N
SMILES:
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1
1
–
1
of
1
results
1
Pipemidic acid, 98%
CAS: 51940-44-4 Molecular Formula: C14H17N5O3 Molecular Weight (g/mol): 303.32 MDL Number: MFCD00057291 InChI Key: JOHZPMXAZQZXHR-UHFFFAOYSA-N PubChem CID: 4831 ChEBI: CHEBI:75250 IUPAC Name: 8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid SMILES: CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1