accessibility menu, dialog, popup

UserName

CAS RN 51934-41-9

H3COOI

CAS RN 51934-41-9

IUPAC Name: ethyl 4-iodobenzoate
Synonyms: ethyl 4-iodobenzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: YCBJOQUNPLTBGG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(I)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)