accessibility menu, dialog, popup

UserName

H3CCH3H3COOOHONHOO

CAS RN 51644-83-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: GLMODRZPPBZPPB-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results