accessibility menu, dialog, popup

UserName

CAS RN 51632-16-7

BrO

CAS RN 51632-16-7

IUPAC Name: 1-(bromomethyl)-3-phenoxybenzene
Synonyms: 1-(bromomethyl)-3-phenoxybenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: UJUNUASMYSTBSK-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(OC2=CC=CC=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results