CAS RN 51549-39-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
CAS: 51549-39-4 Molecular Formula: C23H31N5O4Si Molecular Weight (g/mol): 469.62 MDL Number: MFCD04972281 InChI Key: BJZZQJFYKQPUGO-TWAUYLRJNA-N IUPAC Name: N-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide SMILES: CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12