CAS RN 515-03-7
CAS RN 515-03-7
IUPAC Name:
(1R,2R,8aS)-1-[(3R)-3-hydroxy-3-methylpent-4-en-1-yl]-2,5,5,8a-tetramethyl-decahydronaphthalen-2-ol
Synonyms:
sclareol
Molecular Weight (g/mol):
308.51
Molecular Formula:
C20H36O2
InChi Key:
XVULBTBTFGYVRC-FFADBYAMSA-N
SMILES:
C[C@@](O)(CC[C@H]1[C@](C)(O)CCC2C(C)(C)CCC[C@]12C)C=C
1
–
4
of
4
results
1
Thermo Scientific Chemicals Sclareol
CAS: 515-03-7 Molecular Formula: C20H36O2
2
Sclareol (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sclareol (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Sclareol (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.