accessibility menu, dialog, popup

UserName

CAS RN 51333-90-5

ONCl

CAS RN 51333-90-5

IUPAC Name: 1-(2-chloroethyl)pyrrolidin-2-one
Synonyms: 1-(2-chloroethyl)pyrrolidin-2-one
Molecular Weight (g/mol): 147.6
Molecular Formula: C6H10ClNO
InChi Key: CWLYHDWHNFLUEI-UHFFFAOYSA-N
SMILES: ClCCN1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results