CAS RN 51227-30-6
CAS RN 51227-30-6
IUPAC Name:
1-(4-methylpyridin-3-yl)ethan-1-one
Synonyms:
1-(4-methylpyridin-3-yl)ethanone
Molecular Weight (g/mol):
135.17
Molecular Formula:
C8H9NO
InChi Key:
LCEINQHGZAFADC-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(C)C=CN=C1
1
–
1
of
1
results
1
3-Acetyl-4-methylpyridine, 96%
CAS: 51227-30-6 Molecular Formula: C8H9NO Molecular Weight (g/mol): 135.166 MDL Number: MFCD08447082 InChI Key: LCEINQHGZAFADC-UHFFFAOYSA-N Synonym: 1-4-methylpyridin-3-yl ethanone,3-acetyl-4-methylpyridine,1-4-methyl-3-pyridinyl ethanone,1-4-methyl-3-pyridinyl-ethanone,1-4-methylpyridin-3-yl ethan-1-one,1-4-methyl-pyridin-3-yl-ethanone,ethanone, 1-4-methyl-3-pyridinyl,4-methyl-3-acetylpyridine,3-acetyl-4-picoline,methyl 4-picolyl ketone PubChem CID: 10942481 IUPAC Name: 1-(4-methylpyridin-3-yl)ethanone SMILES: CC1=C(C=NC=C1)C(=O)C