accessibility menu, dialog, popup

UserName

CAS RN 5122-20-3

CH3IH3CCH3H3CCH3

CAS RN 5122-20-3

IUPAC Name: 5-tert-butyl-2-iodo-1,3-dimethylbenzene
Synonyms: 5-tert-butyl-2-iodo-1,3-dimethylbenzene
Molecular Weight (g/mol): 288.17
Molecular Formula: C12H17I
InChi Key: SCKWYPTZVFBKKW-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1I)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results