accessibility menu, dialog, popup

UserName

CAS RN 5096-73-1

IUPAC Name: 6-chloropyridazine-3-carboxylic acid
Synonyms: 6-chloropyridazine-3-carboxylic acid
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: HHGZQZULOHYEOH-UHFFFAOYSA-N
SMILES: OC(=O)C1=NN=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results