accessibility menu, dialog, popup

UserName

HClCH3OOOONCH3H3CCH3

CAS RN 5080-50-2

IUPAC Name: hydrogen 3-(acetyloxy)-4-(trimethylazaniumyl)butanoate chloride
Synonyms: acetyl carnitine hcl
Molecular Weight (g/mol): 239.7
Molecular Formula: C9H18ClNO4
InChi Key: JATPLOXBFFRHDN-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(=O)OC(CC([O-])=O)C[N+](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results