accessibility menu, dialog, popup

UserName

CAS RN 507-20-0

CH3CH3CH3Cl

CAS RN 507-20-0

IUPAC Name: 2-chloro-2-methylpropane
Synonyms: tert-butyl chloride T-butyl chloride
Molecular Weight (g/mol): 92.57
Molecular Formula: C4H9Cl
InChi Key: NBRKLOOSMBRFMH-UHFFFAOYSA-N
SMILES: CC(C)(C)Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)