accessibility menu, dialog, popup

UserName

CAS RN 505-52-2

HOOOHO

CAS RN 505-52-2

IUPAC Name: tridecanedioic acid
Synonyms: brassylic acid monosodium salt tridecanedioic acid disodium salt tridecanedioic acid
Molecular Weight (g/mol): 244.33
Molecular Formula: C13H24O4
InChi Key: DXNCZXXFRKPEPY-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)