accessibility menu, dialog, popup

UserName

CAS RN 503-17-3

CH3H3C

CAS RN 503-17-3

IUPAC Name: but-2-yne
Synonyms: 2-butyne
Molecular Weight (g/mol): 54.09
Molecular Formula: C4H6
InChi Key: XNMQEEKYCVKGBD-UHFFFAOYSA-N
SMILES: CC#CC
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)