accessibility menu, dialog, popup

UserName

H3CCH3CH3CH3H3CCH3SOONN

CAS RN 50257-40-4

IUPAC Name: 1-[2,4,6-tris(propan-2-yl)benzenesulfonyl]-1H-imidazole
Synonyms: 1-(2,4,6-triisopropylbenzenesulfonyl)imidazole
Molecular Weight (g/mol): 334.48
Molecular Formula: C18H26N2O2S
InChi Key: AGGRGODMKWLSDE-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results