accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHOOHOCH3

CAS RN 500788-86-3

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-3-(3-methoxyphenyl)propanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-3-(3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: DNXONWHJJNOKEU-UHFFFAOYNA-N
SMILES: COC1=CC=CC(=C1)C(CC(O)=O)NC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results