accessibility menu, dialog, popup

UserName

CAS RN 5000-65-7

CH3OOBr

CAS RN 5000-65-7

IUPAC Name: 2-bromo-1-(3-methoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-methoxyphenyl)ethanone
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: IOOHBIFQNQQUFI-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)