CAS RN 50-56-6
CAS RN 50-56-6
IUPAC Name:
(2S)-2-{[(2S)-1-[(4R,7S,10S,13S,16S,19R)-19-amino-13-[(2S)-butan-2-yl]-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl]formamido}-N-(carbamoylmethyl)-4-methylpentanamide
Synonyms:
oxytocin ocytocin oxitocina endopituitrina oxytocine syntocinon OXT piton S pitocin (parke-davis) orasthin
Molecular Weight (g/mol):
1007.19
Molecular Formula:
C43H66N12O12S2
InChi Key:
XNOPRXBHLZRZKH-DSZYJQQASA-N
SMILES:
CC[C@H](C)[C@@H]1NC(=O)[C@H](CC2=CC=C(O)C=C2)NC(=O)[C@@H](N)CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCC(N)=O