accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFFFFFFOOH2C

CAS RN 4998-38-3

IUPAC Name: 1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11-icosafluoroundecyl prop-2-enoate
Synonyms: 1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11-icosafluoroundecyl prop-2-enoate
Molecular Weight (g/mol): 586.17
Molecular Formula: C14H6F20O2
InChi Key: HBFWXGZKVGSZKO-UHFFFAOYNA-N
SMILES: FCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)OC(=O)C=C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results