accessibility menu, dialog, popup

UserName

CAS RN 4995-91-9

NiH3COOH3COO

CAS RN 4995-91-9

IUPAC Name: nickel(2+) dioctanoate
Synonyms: nickel(II) octanoate
Molecular Weight (g/mol): 345.11
Molecular Formula: C16H30NiO4
InChi Key: DVTHIMLUHWEZOM-UHFFFAOYSA-L
SMILES: [Ni++].CCCCCCCC([O-])=O.CCCCCCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results