accessibility menu, dialog, popup

UserName

CAS RN 49805-30-3

NHO

CAS RN 49805-30-3

IUPAC Name: 2-azabicyclo[2.2.1]hept-5-en-3-one
Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: DDUFYKNOXPZZIW-UHFFFAOYNA-N
SMILES: O=C1NC2CC1C=C2
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)