CAS RN 498-74-8
CAS RN 498-74-8
IUPAC Name:
3-amino-4-methoxybenzene-1-sulfonyl fluoride
Synonyms:
3-amino-4-methoxybenzenesulfonyl fluoride
Molecular Weight (g/mol):
205.2
Molecular Formula:
C7H8FNO3S
InChi Key:
SESLQGPHIQXYGF-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1N)S(F)(=O)=O
1
–
1
of
1
results
1
4-Methoxymetanilyl fluoride, 97%, Thermo Scientific™
CAS: 498-74-8 Molecular Formula: C7H8FNO3S Molecular Weight (g/mol): 205.2 MDL Number: MFCD00007417 InChI Key: SESLQGPHIQXYGF-UHFFFAOYSA-N Synonym: 4-methoxymetanilyl fluoride,3-amino-4-methoxyphenylsulphonyl fluoride,3-amino-4-methoxy benzene sulphonylfluoride,3-amino-4-methoxyphenyl fluorosulfone,3-amino-4-methoxy-benzenesulfonyl fluoride,3-amino-4-methoxybenzenesulphonyl fluoride,3-azanyl-4-methoxy-benzenesulfonyl fluoride,benzenesulfonylfluoride, 3-amino-4-methoxy,3-amino-4-methoxybenzene-1-sulfonyl fluoride,benzenesulfonyl fluoride, 3-amino-4-methoxy PubChem CID: 96952 IUPAC Name: 3-amino-4-methoxybenzenesulfonyl fluoride SMILES: COC1=C(C=C(C=C1)S(=O)(=O)F)N