accessibility menu, dialog, popup

UserName

CAS RN 498-62-4

IUPAC Name: thiophene-3-carbaldehyde
Synonyms: 3-thiophenecarboxaldehyde
Molecular Weight (g/mol): 112.15
Molecular Formula: C5H4OS
InChi Key: RBIGKSZIQCTIJF-UHFFFAOYSA-N
SMILES: O=CC1=CSC=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)