accessibility menu, dialog, popup

UserName

CAS RN 498-36-2

NaH3CCH3(S)OHOO

CAS RN 498-36-2

IUPAC Name: sodium (2S)-2-hydroxy-4-methylpentanoate
Synonyms: 2-hydroxyisocaproic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C6H11NaO3
InChi Key: WMPVOYIJOLKBNA-JEDNCBNOSA-M
SMILES: [Na+].CC(C)C[C@H](O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results