accessibility menu, dialog, popup

UserName

CAS RN 497-18-7

NH2NHONHH2N

CAS RN 497-18-7

IUPAC Name: 1,3-diaminourea
Synonyms: carbohydrazide carbonic dihydrazide carbonodihydrazide carbonyl dihydrazine hydrazinecarboxamide carbonohydrazid cabazide hydrazide hydrazinecarboxylic acid carbodihydrazide
Molecular Weight (g/mol): 90.09
Molecular Formula: CH6N4O
InChi Key: XEVRDFDBXJMZFG-UHFFFAOYSA-N
SMILES: NNC(=O)NN
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)