accessibility menu, dialog, popup

UserName

NIN

CAS RN 490039-72-0

IUPAC Name: 4-iodopyridine-3-carbonitrile
Synonyms: 4-iodopyridine-3-carbonitrile
Molecular Weight (g/mol): 230.01
Molecular Formula: C6H3IN2
InChi Key: CJKHIIQONPNAIP-UHFFFAOYSA-N
SMILES: IC1=C(C=NC=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)