accessibility menu, dialog, popup

UserName

CAS RN 4900-30-5

H2CCH2

CAS RN 4900-30-5

IUPAC Name: nona-1,8-diene
Synonyms: 1,8-nonadiene
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: VJHGSLHHMIELQD-UHFFFAOYSA-N
SMILES: C=CCCCCCC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results