accessibility menu, dialog, popup

UserName

BrHNN

CAS RN 4887-88-1

IUPAC Name: 6-bromo-1H-1,3-benzodiazole
Synonyms: 5-bromo-3H-1,3-benzodiazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: GEDVWGDBMPJNEV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)