accessibility menu, dialog, popup

UserName

CAS RN 486414-86-2

IUPAC Name: 1-(1-methyl-1H-imidazol-5-yl)methanamine
Synonyms: 1-(3-methylimidazol-4-yl)methanamine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: PYAQTQXFMQWCHQ-UHFFFAOYSA-N
SMILES: CN1C=NC=C1CN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results