accessibility menu, dialog, popup

UserName

CAS RN 481681-02-1

BFOHHOOH

CAS RN 481681-02-1

IUPAC Name: [4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
Synonyms: 4-fluoro-3-(hydroxymethyl)phenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: PWMOQHMTXJYUGE-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results