accessibility menu, dialog, popup

UserName

CAS RN 480-16-0

IUPAC Name: 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
Synonyms: bois d,arc morin (flavonol) morin bois d'arc aurantica C.I. natural yellow 11 al-morin C.I. natural yellow 8 2'-hydroxypelargidenolon 1522 zlut prirodni 11
Molecular Weight (g/mol): 302.24
Molecular Formula: C15H10O7
InChi Key: YXOLAZRVSSWPPT-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(O)=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results