accessibility menu, dialog, popup

UserName

CAS RN 4798-59-8

CH3HOH3C(E)

CAS RN 4798-59-8

IUPAC Name: (2E)-hept-2-en-4-ol
Synonyms: (2E)-hept-2-en-4-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: DODCYMXUZOEOQF-HWKANZRONA-N
SMILES: CCCC(O)\C=C\C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results