accessibility menu, dialog, popup

UserName

CAS RN 4724-48-5

IUPAC Name: octylphosphonic acid
Synonyms: octylphosphonic acid
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H19O3P
InChi Key: NJGCRMAPOWGWMW-UHFFFAOYSA-N
SMILES: CCCCCCCCP(O)(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results