accessibility menu, dialog, popup

UserName

CAS RN 4703-15-5

IUPAC Name: N-(2,6-dimethylphenyl)-4-methylbenzene-1-sulfonamide
Synonyms: N-(2,6-dimethylphenyl)-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 275.37
Molecular Formula: C15H17NO2S
InChi Key: SEYRJIDJWCOIBS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results