accessibility menu, dialog, popup

UserName

HOOClN

CAS RN 4684-94-0

IUPAC Name: 6-chloropyridine-2-carboxylic acid
Synonyms: 6-chloropicolinic acid
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: ZLKMOIHCHCMSFW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)