accessibility menu, dialog, popup

UserName

CAS RN 4671-82-3

CH3(S)POOOPOOO

CAS RN 4671-82-3

IUPAC Name: [(2S)-1-phosphonatopropan-2-yl]phosphonate
Synonyms: (2S)-1-phosphonatopropan-2-ylphosphonate
Molecular Weight (g/mol): 200.03
Molecular Formula: C3H6O6P2
InChi Key: DZQCAOQMXPROIJ-VKHMYHEASA-J
SMILES: C[C@@H](CP([O-])([O-])=O)P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results