accessibility menu, dialog, popup

UserName

CAS RN 4617-33-8

HOOO

CAS RN 4617-33-8

IUPAC Name: 15-hydroxypentadecanoate
Synonyms: 15-hydroxypentadecanoate
Molecular Weight (g/mol): 257.4
Molecular Formula: C15H29O3
InChi Key: BZUNJUAMQZRJIP-UHFFFAOYSA-M
SMILES: OCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results