accessibility menu, dialog, popup

UserName

CAS RN 461-97-2

CH3FCH3

CAS RN 461-97-2

IUPAC Name: 1-fluoro-3,5-dimethylbenzene
Synonyms: 1-fluoro-3,5-dimethylbenzene
Molecular Weight (g/mol): 124.16
Molecular Formula: C8H9F
InChi Key: RCWIWNUVHNAUQC-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results