accessibility menu, dialog, popup

UserName

CAS RN 4605-14-5

IUPAC Name: (3-azaniumylpropyl)({3-[(3-azaniumylpropyl)azaniumyl]propyl})azanium
Synonyms: 3,3,3-tetraminium(4+)
Molecular Weight (g/mol): 192.35
Molecular Formula: C9H28N4
InChi Key: ZAXCZCOUDLENMH-UHFFFAOYSA-R
SMILES: [NH3+]CCC[NH2+]CCC[NH2+]CCC[NH3+]
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)