accessibility menu, dialog, popup

UserName

CAS RN 456-55-3

FFFO

CAS RN 456-55-3

IUPAC Name: (trifluoromethoxy)benzene
Synonyms: trifluoromethoxybenzene
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: GQHWSLKNULCZGI-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results