accessibility menu, dialog, popup

UserName

CAS RN 456-41-7

FBr

CAS RN 456-41-7

IUPAC Name: 1-(bromomethyl)-3-fluorobenzene
Synonyms: 1-(bromomethyl)-3-fluorobenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: SCBZBMXPJYMXRC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CBr)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)