accessibility menu, dialog, popup

UserName

BrBr

CAS RN 4549-32-0

IUPAC Name: 1,8-dibromooctane
Synonyms: octane, 1,8-dibromo-
Molecular Weight (g/mol): 272.02
Molecular Formula: C8H16Br2
InChi Key: DKEGCUDAFWNSSO-UHFFFAOYSA-N
SMILES: BrCCCCCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)