accessibility menu, dialog, popup

UserName

CAS RN 454703-23-2

CH3NHNOOCH3H3CCH3

CAS RN 454703-23-2

IUPAC Name: tert-butyl 3-(ethylamino)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(ethylamino)azetidine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: ZXVCULTZOWFSRS-UHFFFAOYSA-N
SMILES: CCNC1CN(C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)