accessibility menu, dialog, popup

UserName

CAS RN 4546-55-8

N(R)N(R)O(S)(R)NNOHNHHOHOO

CAS RN 4546-55-8

IUPAC Name: N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
Synonyms: adenosine, N-benzoyl-
Molecular Weight (g/mol): 371.35
Molecular Formula: C17H17N5O5
InChi Key: NZDWTKFDAUOODA-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results