accessibility menu, dialog, popup

UserName

CAS RN 4526-07-2

CH3SiCH3CH3SiH3CCH3CH3

CAS RN 4526-07-2

IUPAC Name: trimethyl[4-(trimethylsilyl)buta-1,3-diyn-1-yl]silane
Synonyms: trimethyl[4-(trimethylsilyl)buta-1,3-diyn-1-yl]silane
Molecular Weight (g/mol): 194.42
Molecular Formula: C10H18Si2
InChi Key: LBNVCJHJRYJVPK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results