accessibility menu, dialog, popup

UserName

H3COFBr

CAS RN 450-88-4

IUPAC Name: 1-bromo-4-fluoro-2-methoxybenzene
Synonyms: 1-bromo-4-fluoro-2-methoxybenzene
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: KGYXKRGMSUHYCY-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results