CAS RN 449758-17-2
IUPAC Name:
1-(oxan-2-yl)-1H-pyrazole
Synonyms:
1-(oxan-2-yl)pyrazole
Molecular Weight (g/mol):
152.2
Molecular Formula:
C8H12N2O
InChi Key:
IMZWSOSYNFVECD-UHFFFAOYNA-N
SMILES:
C1CCC(OC1)N1C=CC=N1
1
–
1
of
1
results
1
1-(2-Tetrahydropyranyl)-1H-pyrazole, 98%, Thermo Scientific™
CAS: 449758-17-2 Molecular Formula: C8H12N2O Molecular Weight (g/mol): 152.197 MDL Number: MFCD11044692 InChI Key: IMZWSOSYNFVECD-UHFFFAOYSA-N Synonym: 1-2-tetrahydropyranyl-1h-pyrazole,1-tetrahydro-2h-pyran-2-yl-1h-pyrazole,1-oxan-2-yl-1h-pyrazole,1-oxan-2-yl pyrazole,1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl,1-2-tetrahydropyranyl pyrazole,1-tetrahydro-pyran-2-yl-1h-pyrazole,1-2-oxanyl pyrazole,ksc588k5l,1-tetrahydropyran-2-ylpyrazole PubChem CID: 21866310 IUPAC Name: 1-(oxan-2-yl)pyrazole SMILES: C1CCOC(C1)N2C=CC=N2